missing translation for 'onlineSavingsMsg'
Learn More

8-Anilino-1-naphthalenesulfonic Acid Magnesium(II) Salt Hydrate 99.0+%, TCI America™

Catalog No. A048425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A048425G Supplier TCI America Supplier No. A048425G

Chemical Identifiers

CAS 18108-68-4
Molecular Formula C32H24MgN2O6S2
Molecular Weight (g/mol) 620.977
MDL Number MFCD00065353
InChI Key IRKGXNAZPUOEMW-UHFFFAOYSA-L
Synonym unii-a9jt6a24ev, ansa magnesium salt, ans-mg, 1-naphthalenesulfonic acid, 8-phenylamino-, magnesium salt 2:1, 1,8-ans magnesium salt, a9jt6a24ev, magnesium 1-anilino-8-naphthalenesulfonate, 8-anilino-1-naphthalenesulfonic acid magnesium salt, magnesium 8-anilinonaphthalene-1-sulfonate, magnesium 8-phenylamino naphthalene-1-sulphonate
PubChem CID 87458
IUPAC Name magnesium;8-anilinonaphthalene-1-sulfonate
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)[O-].C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)[O-].[Mg+2]

Specifications

Quantity 25 g
Formula Weight 620.98
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic Acid Magnesium(II) Salt Hydrate