Learn More
Sklar Instruments Scissors, Merit Metzenbaum Dissecting Scissors, Sklar Blunt/Blunt, 7 in., Curved, Smooth, Stainless Steel, EA.
Numéro de catalogue. 501094122
Encompass
Scissors, Merit Metzenbaum Dissecting Scissors, Sklar, Blunt/Blunt, 7 in., Curved, Smooth, Stainless Steel, EA. Merit Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting fine tissue. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
Numéro de catalogue. 501094122 Fournisseur Sklar Instruments Code fournisseur 97340
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Scissors, Merit Metzenbaum Dissecting Scissors, Sklar, Blunt/Blunt, 7 in., Curved, Smooth, Stainless Steel, EA. Merit Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting fine tissue. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.