missing translation for 'onlineSavingsMsg'
Learn More

7H-Dibenzo(c,g)Carbazole, SPEX CertiPrep™

Catalog No. s1197
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S1197 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1197 Supplier SPEX CertiPrep Group LLC Supplier No. S1197
Only null left

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
7H-Dibenzo(c,g)carbazole194-59-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 194-59-2 , 75-09-2
Molecular Formula C20H13N
Molecular Weight (g/mol) 267.331
InChI Key STJXCDGCXVZHDU-UHFFFAOYSA-N
PubChem CID 9134
ChEBI CHEBI:82312
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=C(N3)C=CC5=CC=CC=C54

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Chemical Name or Material 7H-Dibenzo(c,g)carbazole
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only