missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Microliter Syringes: N Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.

Supplier:  Hamilton™ HAM80383

 Afficher plus de versions de ce produit

Numéro de catalogue. 14816208B


Il en reste null

Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Spécifications

Identifiants chimiques

Not Autoclavable
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe

Spécifications

Not Autoclavable
26s G
0.005 in.
0.0283 in.
0.485 mm
0.72 mm
2 in.
51 mm
2000 psi
50°C
701
Ea.
Blunt
Stainless Steel (Plunger)
0.019 in.
0.13 mm
0.305 in.
7.75 mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe
122°F
50°F
3
10 μL
Cemented
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions