missing translation for 'onlineSavingsMsg'
Learn More

7-Octyn-1-ol 98.0+%, TCI America™

Catalog No. O04455ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04455ML 5 mL
O04451ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04455ML Supplier TCI America Supplier No. O04455ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871-91-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD01632137
InChI Key ATCNYMVVGBLQMQ-UHFFFAOYSA-N
PubChem CID 70100
IUPAC Name oct-7-yn-1-ol
SMILES C#CCCCCCCO

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 mL
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Octyn-1-ol