missing translation for 'onlineSavingsMsg'
Learn More

7-Methylquinoline (contains 25% 5-form at maximum) 75.0+%, TCI America™

Catalog No. M04175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04175G 5 g
M041725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04175G Supplier TCI America Supplier No. M04175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 612-60-2
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006805
InChI Key KDYVCOSVYOSHOL-UHFFFAOYSA-N
Synonym quinoline, 7-methyl, m-toluquinoline, unii-eow995k43c, 7-methyl quinoline, ccris 2895, quinolinol,7-methyl, 7-methyl-quinoline, 7-methylchinolin, 7-methylchinolin;, pubchem5811
PubChem CID 11927
IUPAC Name 7-methylquinoline
SMILES CC1=CC2=C(C=CC=N2)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Methylquinoline (contains 25% 5-form at maximum)
EINECSNumber (5)-0796
RTECSNumber VC0560000
TSCA Yes