missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-8-nitroquinoline 98.0+%, TCI America™

Catalog No. M18645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18645G Supplier TCI America Supplier No. M18645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7471-63-8
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
InChI Key ZZDTVYJYMRSNQL-UHFFFAOYSA-N
Synonym quinoline, 7-methyl-8-nitro, 8-nitro-7-methylquinoline, 7-methyl-8-nitro-quinoline, acmc-1bcox, 7-methyl 8-nitroquinoline
PubChem CID 81981
IUPAC Name 7-methyl-8-nitroquinoline
SMILES CC1=C(C2=C(C=CC=N2)C=C1)[N+](=O)[O-]

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methyl-8-nitroquinoline