missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyflavone 98.0+%, TCI America™

Numéro de catalogue. M14235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M14235G 5 g
M14231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M14235G Fournisseur TCI America Code fournisseur M14235G
Il en reste null

Identifiants chimiques

CAS 22395-22-8
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00017462
InChI Key QKNDCRMJDZLFEG-UHFFFAOYSA-N
Synonym 7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one
PubChem CID 466268
IUPAC Name 7-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3

Spécifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
Formula Weight 252.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxyflavone