missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde 97.0+%, TCI America™
Supplier: TCI America I09281G
| Quantity | 1 g |
|---|
Chemical Identifiers
| C16H18O | |
| MFCD00275723 | |
| 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde | |
| 3,8-dimethyl-5-(propan-2-yl)azulene-1-carbaldehyde |
Specifications
| 7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde | |
| 83°C | |
| C16H18O | |
| MFCD00275723 | |
| CDVRGGMPPUFSFH-UHFFFAOYSA-N | |
| 3,8-dimethyl-5-(propan-2-yl)azulene-1-carbaldehyde | |
| 639851 | |
| ≥97.0% (GC) |
| 3331-47-3 | |
| Black-Purple | |
| 1 g | |
| 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde | |
| CC(C)C1=CC=C(C)C2=C(C=O)C=C(C)C2=C1 | |
| 226.32 | |
| 226.32 | |
| Crystalline Powder |
Safety and Handling
TSCA : No