Learn More
7-Hydroxyisoquinoline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1706703
Description
7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.
Solubility
Soluble in methanol and concentrated sulfuric acid.
Notes
Incompatible with oxidizing agents.
Specifications
| 7-Hydroxyisoquinoline | |
| 226°C to 228°C | |
| MFCD00456131 | |
| 113224 | |
| Soluble in methanol and concentrated sulfuric acid. | |
| OC1=CC=C2C=CN=CC2=C1 | |
| 145.16 | |
| 145.16 |
| 7651-83-4 | |
| C9H7NO | |
| 1 g | |
| 7-hydroxyisoquinoline, 7-isoquinolinol, 7-hydroxy-isoquinoline, isoquinolin-7-ol, 7-hydroxy-2-azanaphthalene, pubchem6264, acmc-1bfxl, ksc380o3r, 7-hydroxyisoquinoline 250mg | |
| WCRKBMABEPCYII-UHFFFAOYSA-N | |
| isoquinolin-7-ol | |
| 459767 | |
| 97% |
Chemical Identifiers
| 7651-83-4 | |
| 145.16 | |
| WCRKBMABEPCYII-UHFFFAOYSA-N | |
| 459767 | |
| OC1=CC=C2C=CN=CC2=C1 |
| C9H7NO | |
| MFCD00456131 | |
| 7-hydroxyisoquinoline, 7-isoquinolinol, 7-hydroxy-isoquinoline, isoquinolin-7-ol, 7-hydroxy-2-azanaphthalene, pubchem6264, acmc-1bfxl, ksc380o3r, 7-hydroxyisoquinoline 250mg | |
| isoquinolin-7-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
RUO – Research Use Only