missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyisoquinoline, 97%, Thermo Scientific Chemicals

Catalog No. p-7058471
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17067MD 250 mg
AAL1706703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17067MD Supplier Thermo Scientific Chemicals Supplier No. L17067MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7651-83-4 | Molecular Formula: C9H7NO | Molecular Weight: 145.16 g/mol

7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Hydroxyisoquinoline is used to study its photophysial behavior in polymer matrices such as polymethyl methacrylate, cellulose acetate, polyvinyl alcohol and sulfonated tetrafluoroethylene based fluoropolymer- copolymer. It also used to prepare quinine. Further, it is used as the material for all optical switching due to its excited state proton transfer (ESPT) effect.

Solubility
Soluble in methanol and concentrated sulfuric acid.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7651-83-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00456131
InChI Key WCRKBMABEPCYII-UHFFFAOYSA-N
Synonym 7-hydroxyisoquinoline, 7-isoquinolinol, 7-hydroxy-isoquinoline, isoquinolin-7-ol, 7-hydroxy-2-azanaphthalene, pubchem6264, acmc-1bfxl, ksc380o3r, 7-hydroxyisoquinoline 250mg
PubChem CID 459767
IUPAC Name isoquinolin-7-ol
SMILES OC1=CC=C2C=CN=CC2=C1

Specifications

Melting Point 226°C to 228°C
Quantity 250 mg
Beilstein 113224
Solubility Information Soluble in methanol and concentrated sulfuric acid.
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material 7-Hydroxyisoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only