missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1-indanone 98.0+%, TCI America™

Catalog No. H15261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15261G Supplier TCI America Supplier No. H15261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6968-35-0
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD01548387
InChI Key HFMZPBSZKCDKOR-UHFFFAOYSA-N
Synonym 7-hydroxy-1-indanone, 7-hydroxy-2,3-dihydro-1h-inden-1-one, 7-hydroxyindan-1-one, 1h-inden-1-one,2,3-dihydro-7-hydroxy, 4h-inden-4-one, 1,2-dihydro-3-hydroxy, 7-oxidanyl-2,3-dihydroinden-1-one, 7-hydroxyindanone, acmc-20mvbk, pubchem23254, acmc-209wv8
PubChem CID 248078
IUPAC Name 7-hydroxy-2,3-dihydro-1H-inden-1-one
SMILES OC1=CC=CC2=C1C(=O)CC2

Specifications

Melting Point 112°C
Quantity 1 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-1-indanone