missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzofurazan-4-sulfonamide, 98%

Catalog No. AAB23586MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23586MC 100 mg
AAB23586MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23586MC Supplier Thermo Scientific Chemicals Supplier No. B23586MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91366-65-3 | C6H4FN3O3S | 217.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91366-65-3
Molecular Formula C6H4FN3O3S
Molecular Weight (g/mol) 217.17
MDL Number MFCD00036687
InChI Key XROXHZMRDABMHS-UHFFFAOYSA-N
Synonym abd-f, 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzofurazan-4-sulfonamide, 4-fluoro-7-sulfamoylbenzofurazan, 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonamide, 4-aminosulphonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonamide, pubchem23230, 4-fluoro-7-sulfamoylbenzofurazane, 4-fluoro-7-sulfoamoylbenzofurazan
PubChem CID 122067
IUPAC Name 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(F)C2=NON=C12

Specifications

Melting Point 143°C to 147°C
Quantity 100 mg
Formula Weight 217.17
Percent Purity 98%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only