missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 7-Diethylamino-4-methylcoumarin, 99%

Catalog No. AC114055000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114055000 500 g Plastic bottle
AC114050050 5 g Glass Bottle
AC114051000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114055000 Supplier Thermo Scientific Chemicals Supplier No. 114055000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-44-1 | C14H17NO2 | 231.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-44-1
Molecular Formula C14H17NO2
Molecular Weight (g/mol) 231.29
MDL Number MFCD00006864
InChI Key AFYCEAFSNDLKSX-UHFFFAOYSA-N
Synonym 7-diethylamino-4-methylcoumarin, coumarin 1, 7-diethylamino-4-methyl-2h-chromen-2-one, coumarin 47, coumarin 460, 4-methyl-7-diethylamino coumarin, blancophor aw, blancophor ffg, 4-methyl-7-diethylaminocoumarin, hakkol p
PubChem CID 7050
ChEBI CHEBI:51938
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige to Purple
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous acid solutions,soluble in nearly all common organic solvents,slightly soluble in aliphatic hydrocarbons
Formula Weight 231.29
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Diethylamino-4-methylcoumarin, 99%
Show More Show Less
EINECSNumber 202-068-9
RTECSNumber GN6370000
TSCA TSCA

RUO – Research Use Only