missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Deazaguanine, 90+%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 7-Deazaguanine |
| Fourchette de pourcentages d’essai | 90% min. (HPLC) |
| Emballage | Glass bottle |
| Quantité | 1g |
| Poids de formule | 150.14 |
| Pourcentage de pureté | 90+% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only