missing translation for 'onlineSavingsMsg'
Learn More

7-Chloropyrazolo[1,5-a]pyrimidine, 95%

Catalog No. AC438490050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438490050 5 g Glass bottle
AC438490010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438490050 Supplier Thermo Scientific Chemicals Supplier No. 438490050
Only null left

CAS: 58347-49-2 | C6H4ClN3 | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58347-49-2
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD04035684
InChI Key YCZQHXPIKQHABJ-UHFFFAOYSA-N
Synonym 7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine
PubChem CID 12281647
IUPAC Name 7-chloropyrazolo[1,5-a]pyrimidine
SMILES C1=C2N=CC=C(N2N=C1)Cl

Specifications

Melting Point 98.0°C to 102.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 153.57
Percent Purity 95%
Physical Form Powder
Assay 95%
Chemical Name or Material 7-Chloropyrazolo[1,5-a]pyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only