missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 244°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 281.67 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid |
Safety and Handling
| RTECSNumber | VB1993795 |
| TSCA | No |