missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%

Catalog No. AAH63456MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63456MD 250 mg
AAH6345603 1 g
AAH6345606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63456MD Supplier Thermo Scientific Chemicals Supplier No. H63456MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73075-45-3 | C9H11Cl2N | 204.094 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 73075-45-3
Molecular Formula C9H11Cl2N
Molecular Weight (g/mol) 204.094
MDL Number MFCD08461076
InChI Key OGIAIXMUSSACDB-UHFFFAOYSA-N
Synonym 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 7-chloro-1,2,3,4-tetrahydro-, hydrochloride, 7-chloro-1,2,3,4-tetrahydro-isoquinoline hcl, 7-chhloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochlor, 7-chloro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 12595070
IUPAC Name 7-chloro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)Cl.Cl

Specifications

Quantity 250 mg
Formula Weight 204.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only