missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoisatina 97,0+%, TCI America™

Catalog No. B39745G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B39745G 5 g
B39741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39745G Supplier TCI America Supplier No. B39745G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20780-74-9
Formule moléculaire C8H4BrNO2
Poids moléculaire (g/mol) 226.029
Numéro MDL MFCD00774354
Clé InChI OCVKSIWBTJCXPV-UHFFFAOYSA-N
Synonyme 7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin
PubChem CID 2302353
Nom de l’IUPAC 7-bromo-1H-indole-2,3-dione
SOURIRES C1=CC2=C(C(=C1)Br)NC(=O)C2=O

Specifications

Nom ou substance chimique 7-Bromoisatin
Point de fusion 200 °C
Quantité 5 g
Poids de formule 226.03
Pourcentage de pureté ≥97,0% (GC)
Forme physique Poudre cristalline