missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine, 95%, Thermo Scientific™

Catalog No. CC13510EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13510EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13510EA Supplier Thermo Scientific Chemicals Supplier No. CC13510EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154264-95-6
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.09
MDL Number MFCD02681913
InChI Key MQMFOFZKZBLSAB-UHFFFAOYSA-N
Synonym 7-bromo-4-methyl-3,4-dihydro-2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-benzo e 1,4-oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine
PubChem CID 2776405
IUPAC Name 7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(Br)=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 228.09
Percent Purity 95%
Chemical Name or Material 7-Bromo-4-methyl-3, 4-dihydro-2H-1, 4-benzoxazine
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.