missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 7,8-Dihydroxyflavone

Catalog No. 382610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
382610 10 mg
382650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 382610 Supplier Tocris Bioscience™ Supplier No. 3826/10*

TrkB agonist

Tyrosine kinase receptor B (TrkB) agonist that binds to the extracellular domain of the receptor (Kd = 320 nM). Inhibits glutamate-triggered apoptosis in hippocampal neurons in vitro and in vivo. Exhibits neuroprotective effects in an HD mouse model.

Chemical Identifiers

CAS 38183-03-8
Molecular Formula C15H10O4

Specifications

Quantity 10 mg
Formula Weight 254.24
Purity >98%
Molecular Weight (g/mol) 254.24
Chemical Name or Material 7, 8-Dihydroxyflavone
Recommended Storage Store at +4°C