accessibility menu, dialog, popup

UserName

CAS RN 38183-03-8

IUPAC Name: 7,8-dihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: 7,8-dihydroxyflavone
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: COCYGNDCWFKTMF-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)C=C(OC2=C1O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)