missing translation for 'onlineSavingsMsg'
Learn More

7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine 98.0+%, TCI America™

Catalog No. p-7123833
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4908200MG 200 mg
B49081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4908200MG Supplier TCI America Supplier No. B4908200MG
Only null left

Chemical Identifiers

CAS 1395881-55-6
Molecular Formula C36H20N2O4
Molecular Weight (g/mol) 544.566
InChI Key LQIJYRYGPVWGLO-UHFFFAOYSA-N
Synonym HN-D1
PubChem CID 89981730
SMILES C1=CC=C2C(=C1)N3C4=CC=CC=C4OC5=C3C(=CC(=C5)C6=CC7=C8C(=C6)OC9=CC=CC=C9N8C1=CC=CC=C1O7)O2

Specifications

Melting Point 376°C
Color Yellow
Quantity 200 mg
Formula Weight 544.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7,7′-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine