Learn More
Chemglass Inc 6mm Diameter Quartz Rod, 48in Length
Numéro de catalogue. CGQ080105
Encompass
6mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.
Numéro de catalogue. CGQ080105 Fournisseur Chemglass Inc Code fournisseur CGQ080105
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
6mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.