Scissors, Merit Multi-Cut Utility Scissors, Sklar, 6 in., Blue, Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA. Merit Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. These scissors feature plastic finger ring handles with one ring that is larger than the other