Learn More
YSI™ 6051020 Pro Series ISE/DO Cable
Cable for Pro Plus DO/pH/temperature meters
Supplier: YSI™ 605102010
Description
- Measure any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), dissolved oxygen and temperature
- Cables over 1-meter also come with the 603062 cable management kit
- The 6051020 cables ONLY work with the YSI Pro Plus handheld
- DOES NOT come with the ISE sensor
Order Info
Order probes separately.
Spécifications
| Cable Management kit | |
| 393.7 in. | |
| 10 m |