missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 6000 Series Thermal Transfer Printer Ribbons, Black

For wash and reflow polyimide labels. Solvent, high heat and scratch resistant. Brady™ 6000 Series Thermal Transfer Printer Ribbons are solvent, high heat and scratch resistant and available in variety of sizes.

$50.43 - $427.05

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) Thermal Transfer Printer
Material Polyester
Description Ribbon Series: 6000
View More Specs

Products 9
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity & Availability  
19104623
View Documents Promotion Details
Brady™
BRDR6001
360 ft. 4 in.
Each for $100.00
Only null left
 
19104624
View Documents Promotion Details
Brady™
R6002
984 ft. 3.27 in.
Each for $220.21
Only null left
 
19104626
View Documents Promotion Details
Brady™
R6003
500 ft. 1.57 in.
Each for $50.43
Only null left
 
19104627
View Documents Promotion Details
Brady™
R6004
984 ft. 5.12 in.
Each for $314.42
Only null left
 
19104628
View Documents Promotion Details
Brady™
R6005
500 ft. 2.36 in.
Each for $80.18
Only null left
 
19104629
View Documents Promotion Details
Brady™
R6006
984 ft. 1.57 in.
Each for $100.00
Only null left
 
19104630
View Documents Promotion Details
Brady™
R6007
984 ft. 4.33 in.
Each for $321.75
Only null left
 
19104632
View Documents Promotion Details
Brady™
R6008
984 ft. 6.85 in.
Each for $427.05
Only null left
 
19104633
View Documents Promotion Details
Brady™
R6009
500 ft. 4.33 in.
Each for $138.74
Only null left
 
Description

Description

  • Formulation: Resin
  • Compatible with Thermal Transfer printers
Specifications

Specifications

Printer Ribbon
Thermal Transfer Printer
Ribbon Series: 6000
Black
Polyester
Videos
SDS
Documents

Documents