missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ 6000 Series Thermal Transfer Printer Ribbons, Black
For wash and reflow polyimide labels. Solvent, high heat and scratch resistant. Brady™ 6000 Series Thermal Transfer Printer Ribbons are solvent, high heat and scratch resistant and available in variety of sizes.
Supplier: Brady™ R6004
Description
- Formulation: Resin
- Compatible with Thermal Transfer printers
Chemical Identifiers
| Printer Ribbon | |
| Thermal Transfer Printer | |
| Ribbon Series: 6000 |
| Black | |
| Polyester |
Specifications
| Printer Ribbon | |
| Thermal Transfer Printer | |
| 984 ft. | |
| Ribbon Series: 6000 |
| Black | |
| Polyester | |
| 5.12 in. |