missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 6000 Series Thermal Transfer Printer Ribbons, Black

For wash and reflow polyimide labels. Solvent, high heat and scratch resistant. Brady™ 6000 Series Thermal Transfer Printer Ribbons are solvent, high heat and scratch resistant and available in variety of sizes.

Supplier:  Brady™ R6003

 View more versions of this product

Catalog No. 19104626


Only null left

Description

Description

  • Formulation: Resin
  • Compatible with Thermal Transfer printers
Specifications

Chemical Identifiers

Printer Ribbon
Thermal Transfer Printer
Ribbon Series: 6000
Black
Polyester

Specifications

Printer Ribbon
Thermal Transfer Printer
500 ft.
Ribbon Series: 6000
Black
Polyester
1.57 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions