missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 60 Well HLA Terasaki Plates

Suitable for all applications for serological determination of HLA antigens
Supplier: Greiner Bio-One 653180
Description
- Suitable for all applications for serological determination of HLA antigens
- Manufactured from crystal clear polystyrene
- Stackable
- Improved cell adhesion through physical surface treatment
Spécifications
| Clear | |
| Polystyrene | |
| V | |
| Clear | |
| Rectangular | |
| Tissue Culture | |
| 60 | |
| Lid | |
| 10 | |
| 270/Cs. | |
| 83 mm x 58 mm x 11 mm | |
| 58 mm |
| Closure Included | |
| Not Autoclavable | |
| 10 L (Well) | |
| Polystyrene | |
| Non-sterile | |
| Microtest Plate | |
| HLA testing | |
| 10 pcs/bag | |
| Inner bags of 10 | |
| -20°C to +60°C | |
| 83.3 mm | |
| 11 mm |
Sécurité et manipulation
missing translation for 'shelfLife' : 1 Year