missing translation for 'onlineSavingsMsg'
Learn More

6-Undecanol 98.0+%, TCI America™

Catalog No. U00405G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00405G Supplier TCI America Supplier No. U00405G
Only null left

Chemical Identifiers

CAS 23708-56-7
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00021951
InChI Key YBIXBBGRHOUVBB-UHFFFAOYSA-N
Synonym 6-undecanol, undecanol-6, diamyl carbinol, acmc-1chvn
PubChem CID 32045
IUPAC Name undecan-6-ol
SMILES CCCCCC(O)CCCCC

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 172.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Undecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No