missing translation for 'onlineSavingsMsg'
Learn More

6-Undecanamine 97.0+%, TCI America™

Catalog No. U00945ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00945ML 5 mL
U00941ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00945ML Supplier TCI America Supplier No. U00945ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33788-00-0
Molecular Formula C11H25N
Molecular Weight (g/mol) 171.328
MDL Number MFCD00047924
InChI Key GFBRYGJZWXLRFR-UHFFFAOYSA-N
Synonym 1-Pentylhexylamine, 6-Aminoundecane
PubChem CID 141845
IUPAC Name undecan-6-amine
SMILES CCCCCC(CCCCC)N

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
UN Number 2735
Formula Weight 171.33
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 6-Undecanamine
EINECSNumber (2)-0133&(2)-0176
TSCA No