missing translation for 'onlineSavingsMsg'
Learn More

(6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC54863DA

Quantity 1g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC54863DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

913830-32-7
263.18
FWNBHEVGWIALSL-UHFFFAOYSA-N
24229545
C1=CSC(=C1)C2=NC=C(C=C2)CN.Cl.Cl
C10H12Cl2N2S
MFCD09025878
6-thien-2-ylpyridin-3-yl methylamine dihydrochloride, 6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 1-6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 6-thiophen-2-yl pyrid-3-ylmethylamine dihydrochloride, 5-aminomethyl-2-thien-2-yl pyridine dihydrochloride, 6-2-thienyl-3-pyridyl methylamine, chloride, chloride, 2-5-aminomethyl pyridin-2-yl thiophene dihydrochloride, 3-pyridinemethanamine,6-2-thienyl-,hydrochloride 1:2, 1-6-thiophen-2-yl pyridin-3-yl methanamine-hydrogen chloride 1/2
(6-thiophen-2-ylpyridin-3-yl)methanamine;dihydrochloride

Spécifications

(6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride
89.5
90%
C10H12Cl2N2S
MFCD09025878
FWNBHEVGWIALSL-UHFFFAOYSA-N
(6-thiophen-2-ylpyridin-3-yl)methanamine;dihydrochloride
24229545
90%
913830-32-7
100.0
Amber Glass Bottle
1g
6-thien-2-ylpyridin-3-yl methylamine dihydrochloride, 6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 1-6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 6-thiophen-2-yl pyrid-3-ylmethylamine dihydrochloride, 5-aminomethyl-2-thien-2-yl pyridine dihydrochloride, 6-2-thienyl-3-pyridyl methylamine, chloride, chloride, 2-5-aminomethyl pyridin-2-yl thiophene dihydrochloride, 3-pyridinemethanamine,6-2-thienyl-,hydrochloride 1:2, 1-6-thiophen-2-yl pyridin-3-yl methanamine-hydrogen chloride 1/2
C1=CSC(=C1)C2=NC=C(C=C2)CN.Cl.Cl
263.18
263.19
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

FDS
Documents

Documents