missing translation for 'onlineSavingsMsg'
Learn More

6-tert-Butyl-m-cresol 97.0+%, TCI America™

Numéro de catalogue. B0718500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B0718500G 500 g
B071825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B0718500G Fournisseur TCI America Code fournisseur B0718500G
Il en reste null

Identifiants chimiques

CAS 88-60-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002308
InChI Key XOUQAVYLRNOXDO-UHFFFAOYSA-N
Synonym 6-tert-butyl-m-cresol, 6-tert-butyl-3-methylphenol, 2-tert-butyl-5-methylphenol, m-cresol, 6-tert-butyl, 3-methyl-6-tert-butylphenol, phenol, 2-1,1-dimethylethyl-5-methyl, 5-methyl-2-tert-butylphenol, unii-uxv32mj2ca, 2-tert-butyl-5-methyl-phenol, 6-t-butyl-m-cresol
PubChem CID 6937
IUPAC Name 2-tert-butyl-5-methylphenol
SMILES CC1=CC(=C(C=C1)C(C)(C)C)O

Spécifications

Boiling Point 244°C
Quantity 500 g
UN Number 2430
Formula Weight 164.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-tert-Butyl-m-cresol
EINECSNumber (3)-0521
RTECSNumber SK1578180
TSCA Yes