missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinylmethanol, 97%, Thermo Scientific™

Numéro de catalogue. p-3075862
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC04509CB 250mg Amber Glass Bottle
CC04509DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC04509CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC04509CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 100516-88-9
Formule moléculaire C10H9NO
Poids moléculaire (g/mol) 159.188
Numéro MDL MFCD03789621
Clé InChI YQEJIIUSNDZIGO-UHFFFAOYSA-N
Synonyme 6-quinolinylmethanol, 6-quinolinemethanol, 6-hydroxymethylquinoline, quinolin-6-yl methanol, 6-quinolylmethanol, pubchem12427, quinolin-6-yl-methanol, 6-hydroxymethyl quinoline, 6-hydroxymethyl-quinoline
PubChem CID 1514385
Nom de l’IUPAC quinolin-6-ylmethanol
SOURIRES C1=CC2=C(C=CC(=C2)CO)N=C1

Spécifications

Nom ou substance chimique 6-Quinolinylmethanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 159.19
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.