missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylnicotinonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 39065-54-8
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD06637420
InChI Key OSRYUVXCTNUGRM-UHFFFAOYSA-N
Synonym 6-phenylnicotinonitrile, 5-cyano-2-phenylpyridine, 3-pyridinecarbonitrile,6-phenyl, 3-pyridinecarbonitrile, 6-phenyl, 6-phenylniocotinonitrile, 2-phenyl-5-cyanopyridine, 3-cyano-6-phenylpyridine, 6-phenyl-3-pyridinecarbonitrile
PubChem CID 12576518
IUPAC Name 6-phenylpyridine-3-carbonitrile
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC21116CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC21116DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

39065-54-8
180.21
OSRYUVXCTNUGRM-UHFFFAOYSA-N
12576518
C1=CC=C(C=C1)C2=NC=C(C=C2)C#N
C12H8N2
MFCD06637420
6-phenylnicotinonitrile, 5-cyano-2-phenylpyridine, 3-pyridinecarbonitrile,6-phenyl, 3-pyridinecarbonitrile, 6-phenyl, 6-phenylniocotinonitrile, 2-phenyl-5-cyanopyridine, 3-cyano-6-phenylpyridine, 6-phenyl-3-pyridinecarbonitrile
6-phenylpyridine-3-carbonitrile

Specifications

39065-54-8
100.0
Amber Glass Bottle
MFCD06637420
OSRYUVXCTNUGRM-UHFFFAOYSA-N
6-phenylpyridine-3-carbonitrile
12576518
97%
96.5
97%
C12H8N2
6-phenylnicotinonitrile, 5-cyano-2-phenylpyridine, 3-pyridinecarbonitrile,6-phenyl, 3-pyridinecarbonitrile, 6-phenyl, 6-phenylniocotinonitrile, 2-phenyl-5-cyanopyridine, 3-cyano-6-phenylpyridine, 6-phenyl-3-pyridinecarbonitrile
C1=CC=C(C=C1)C2=NC=C(C=C2)C#N
180.21
180.21
6-Phenylnicotinonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents