missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-3(2H)-pyridazinone 98.0+%, TCI America™

Numéro de catalogue. P20601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P20601G 1 g
P20605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P20601G Fournisseur TCI America Code fournisseur P20601G
Il en reste null

Identifiants chimiques

CAS 2166-31-6
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD00114953
InChI Key IJUIPRDMWWBTTQ-UHFFFAOYSA-N
Synonym 3-Hydroxy-6-phenylpyridazine
PubChem CID 75106
IUPAC Name 3-phenyl-1H-pyridazin-6-one
SMILES C1=CC=C(C=C1)C2=NNC(=O)C=C2

Spécifications

Melting Point 203°C
Color Yellow
Quantity 1 g
Formula Weight 172.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Phenyl-3(2H)-pyridazinone