missing translation for 'onlineSavingsMsg'
Learn More

6-O-Palmitoyl-L-ascorbic Acid 97.0+%, TCI America™

Catalog No. A054025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A054025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A054025G Supplier TCI America Supplier No. A054025G

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
Synonym ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf
PubChem CID 54680660
IUPAC Name 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 25 g
Formula Weight 414.54
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-O-Palmitoyl-L-ascorbic Acid
EINECSNumber (5)-0063
RTECSNumber CI7671040
TSCA Yes