missing translation for 'onlineSavingsMsg'
Learn More

6-O-Methylguanine (2-Amino-6-Methoxypurine) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013486
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013486 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013486 Supplier Merck Emd Millipore Supplier No. PHR17191G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 20535-83-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H7N5 O
MDL Number MFCD00133009
Melting Point >300°C
Synonym 6-O-Methylguanine; 2-Amino-6-methoxypurine; 6-Methoxyguanine
Type Certified Reference Material
Quantity 1 g
Formula Weight 165.15
Grade Certified Reference Material