missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroveratraldehyde 85.0+%, TCI America™

Catalog No. N05385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05385G 5 g
N053825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05385G Supplier TCI America Supplier No. N05385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20357-25-9
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00007134
InChI Key YWSPWKXREVSQCA-UHFFFAOYSA-N
Synonym 6-nitroveratraldehyde, dmnb, 3,4-dimethoxy-6-nitrobenzaldehyde, 4,5-dimethoxy-2-nitro-benzaldehyde, dna-pk inhibitor, benzaldehyde, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzaldehyde, dna-dependent protein kinase inhibitor, 2-nitro-4,5-bis methyloxy benzaldehyde, pubchem8213
PubChem CID 88505
IUPAC Name 4,5-dimethoxy-2-nitrobenzaldehyde
SMILES COC1=CC(C=O)=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 132°C
Quantity 5 g
Formula Weight 211.17
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroveratraldehyde