missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitroindazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 182°C |
| Quantity | 25 g |
| Formula Weight | 163.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Nitroindazole |
Safety and Handling
| RTECSNumber | NK7962100 |
| TSCA | Yes |