missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrobenzothiazole, 98+%

Catalog No. AAA1063418
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1063418 50 g
AAA1063406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1063418 Supplier Thermo Scientific Chemicals Supplier No. A1063418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2942-06-5 | C7H4N2O2S | 180.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2942-06-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.18
MDL Number MFCD00014569
InChI Key QLUFBCVWKTWKBF-UHFFFAOYSA-N
IUPAC Name 6-nitro-1,3-benzothiazole

Specifications

Melting Point 173°C to 177°C
Quantity 50 g
Beilstein 7824
Synonym 6-nitrobenzothiazole, 6-nitrobenzo d thiazole, benzothiazole, 6-nitro, 6-nitro-benzothiazole, pubchem21845, acmc-1ciqi, #, benzothiazole, 6-nitro-6ci,7ci,8ci,9ci
SMILES C1=CC2=C(C=C1[N+](=O)[O-])SC=N2
PubChem CID 76256
Formula Weight 180.18
Percent Purity ≥98%
Chemical Name or Material 6-Nitrobenzothiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-933-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only