missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitro-m-cresol 95.0+%, TCI America™
Supplier: TCI America M1122500G
Specifications
| 6-Nitro-m-cresol | |
| 56°C | |
| MFCD00007111 | |
| 2446 | |
| NQXUSSVLFOBRSE-UHFFFAOYSA-N | |
| 5-methyl-2-nitrophenol | |
| 12788 | |
| ≥95.0% (T) |
| 700-38-9 | |
| C7H7NO3 | |
| 500 g | |
| 6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso | |
| CC1=CC=C(C(O)=C1)[N+]([O-])=O | |
| 153.14 | |
| 153.14 | |
| Crystalline Powder |
Chemical Identifiers
| 700-38-9 | |
| 153.14 | |
| NQXUSSVLFOBRSE-UHFFFAOYSA-N | |
| 12788 | |
| CC1=CC=C(C(O)=C1)[N+]([O-])=O |
| C7H7NO3 | |
| MFCD00007111 | |
| 6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso | |
| 5-methyl-2-nitrophenol |
Safety and Handling
RTECSNumber : SM1130950
TSCA : No