accessibility menu, dialog, popup

UserName

CH3HONOO

CAS RN 700-38-9

IUPAC Name: 5-methyl-2-nitrophenol
Synonyms: 6-nitro-m-cresol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: NQXUSSVLFOBRSE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)