missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-m-cresol 95.0+%, TCI America™

Catalog No. p-7132551
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M112225G 25 g
M1122500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M112225G Supplier TCI America Supplier No. M112225G
Only null left

Chemical Identifiers

CAS 700-38-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007111
InChI Key NQXUSSVLFOBRSE-UHFFFAOYSA-N
Synonym 6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso
PubChem CID 12788
IUPAC Name 5-methyl-2-nitrophenol
SMILES CC1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

Melting Point 56°C
Quantity 25 g
UN Number 2446
Formula Weight 153.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitro-m-cresol
RTECSNumber SM1130950
TSCA No