missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%

Catalog No. AAH33900MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33900MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH33900MD Supplier Thermo Scientific Chemicals Supplier No. H33900MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28226-22-4 | C8H8N2O3 | 180.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28226-22-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD07698591
InChI Key GZAJZBARYACGSO-UHFFFAOYSA-N
Synonym 6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
PubChem CID 7062225
IUPAC Name 6-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material 6-Nitro-3,4-dihydro-2H-1,4-benzoxazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only