missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC38301CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38301CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC38301CB Supplier Thermo Scientific Chemicals Supplier No. CC38301CB

Chemical Identifiers

CAS 554405-17-3
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
InChI Key SAZSDHZXRMXESN-UHFFFAOYSA-N
Synonym 6-morpholinopyridine-2-carboxylic acid, 6-morpholinopicolinic acid, 6-morpholin-4-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-4-morpholinyl, 6-morpholinopyridine-2-carboxylicacid, 6-morpholin-4-yl picolinic acid, 6-4-morpholinyl-2-pyridinecarboxylic acid
PubChem CID 2388921
IUPAC Name 6-morpholin-4-ylpyridine-2-carboxylic acid
SMILES C1COCCN1C2=CC=CC(=N2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 6-Morpholinopyridine-2-carboxylic acid