missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AC454390010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454390010 Supplier Thermo Scientific Chemicals Supplier No. 454390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610768-32-6 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610768-32-6
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD08669608
InChI Key JEECGGXGHJUYMN-UHFFFAOYSA-N
Synonym 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-picoline-5-boronic acid pinacol ester, 6-methylpyridine-3-boronic acid pinacol ester, 2-picoline-5-boronic acid pinacolate, 2-methylpyridine-5-boronic acid, pinacol ester, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-picoline-5-boronic acidpinacolate, 2-methylpyridine-5-boronic acid pinacolate
PubChem CID 16414272
IUPAC Name 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 219.09
Percent Purity 95%
Chemical Name or Material 6-Methylpyridine-3-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only