missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpyridine-2-carboxylic acid, 95%

Catalog No. AAH2630206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2630206 5 g
AAH2630203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2630206 Supplier Thermo Scientific Chemicals Supplier No. H2630206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 934-60-1 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 934-60-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00023481
InChI Key LTUUGSGSUZRPRV-UHFFFAOYSA-N
Synonym 6-methyl-2-pyridinecarboxylic acid, 6-methylpicolinic acid, 6-methyl-2-picolinic acid, 2-carboxy-6-methylpyridine, 6-methyl-2-picolinicacid, 2-pyridinecarboxylic acid, 6-methyl, 2-picoline-6-carboxylic acid, picolinic acid, 6-methyl, 6-methyl picolinic acid, unii-kb675y352l
PubChem CID 70282
IUPAC Name 6-methylpyridine-2-carboxylic acid
SMILES CC1=CC=CC(=N1)C(O)=O

Specifications

Melting Point 127°C to 131°C
Boiling Point 100°C (4.5 mmHg)
Odor Odorless
Quantity 5 g
Formula Weight 137.14
Percent Purity 95%
Chemical Name or Material 6-Methylpyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-287-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only