missing translation for 'onlineSavingsMsg'
Learn More

6-Methylchromone-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. M27255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27255G 5 g
M27251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27255G Supplier TCI America Supplier No. M27255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5006-44-0
Molecular Formula C11H7O4
Molecular Weight (g/mol) 203.17
MDL Number MFCD00239435
InChI Key RKIDLZFIIVDZNX-UHFFFAOYSA-M
Synonym 6-methylchromone-2-carboxylic acid, 6-methyl-4-oxo-4h-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-1-benzopyran-2-carboxylic acid, maybridge3_003393, 4h-1-benzopyran-2-carboxylicacid, 6-methyl-4-oxo
PubChem CID 688704
IUPAC Name 6-methyl-4-oxo-4H-chromene-2-carboxylate
SMILES CC1=CC=C2OC(=CC(=O)C2=C1)C([O-])=O

Specifications

Melting Point 258°C
Quantity 5 g
Formula Weight 204.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylchromone-2-carboxylic Acid